Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, ≥99%, Spectrophotometric Grade, Solstice

Catalog No. 60048504 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-504 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-504 Supplier Solstice Supplier No. 1548061L

missing translation for 'validMainframeMsg'

Spectrophotometric Grade,≥99%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20°C
Density 0.937g/mL at 25°C
Quantity 1 L
Explosion Limit 1.8% (100°F), 11.5% (160°F)
Flash Point 70°C (158°F)
Ignition Point 914°F
Packaging Glass Bottle
Refractive Index 1.437
Linear Formula CH3CON(CH3)2
Vapor Pressure 4mmHg (38°C), 2mmHg (25°C)
Beilstein 1737614
Boiling Point 164°C to 166°C
Formula Weight 87.12g/mol
Percent Purity ≥99%
Grade Spectrophotometry
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
Hazard Statement P201-P261-P280-P305 + P351 + P338-P308 + P313
Precautionary Statement H312 + H332-H319-H360D
EINECSNumber 204-826-4
RTECSNumber AB7700000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.