Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, Anhydrous, DriSolv™, MilliporeSigma™

Catalog No. MDX15446 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MDX15446 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MDX15446 Supplier MilliporeSigma Supplier No. DX15446

missing translation for 'validMainframeMsg'

Septum-Sealed Bottle

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20°C
Color Colorless
Density 0.94g/cm3 at 20°C
Quantity 1 L
Flash Point 69.85°C (157.7°F)
Ignition Point 489.85°C (913.7°F)
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 166.1°C
Solubility Information Soluble in water
Formula Weight 87.12
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept uprightto prevent leakage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.