Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023062 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
3 x 1.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-3062 20 mL
11-102-3007 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-3062 Supplier MilliporeSigma Supelco Supplier No. RTC00010420ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standard for applications in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

CAS 127-19-5
Quantity 20 mL
Flash Point 64°C (Not applicable)
Molecular Formula C4H9NO
Linear Formula C4H9NO
Synonym N,N-Dimethylacetamide solution
Formula Weight 87.12
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.