Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, CHROMASOLV™ Plus, for HPLC, ≥99.9%, Solstice

Catalog No. 6002240 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-022-40 2.5 L
60-045-744 6 x 1 L
60-048-239 4 x 4 L
60-022-39 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-022-40 Supplier Solstice Supplier No. 2705552.5L
Add to Cart
Add to Cart

CHROMASOLV™ Plus, for HPLC,≥99.9%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20°C
Density 0.937g/mL at 25°C
Quantity 2.5 L
Refractive Index n20/D 1.437
Vapor Pressure 2 mmHg (25°C)
Linear Formula CH3CON(CH3)2
Beilstein 1737614
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
Boiling Point 164.5°C to 166°C
Formula Weight 87.12g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Plus
Chemical Name or Material N,N-Dimethylacetamide, for HPLC
Show More Show Less
Hazard Statement P201-P261-P280-P302 + P352 + P312-P304 + P340 + P312-P308 + P313
Precautionary Statement H312 + H332-H319-H360D
EINECSNumber 204-826-4
RTECSNumber AB7700000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.