Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacetamide, CHROMASOLV™ Plus, for HPLC, ≥99.9%, Solstice

Catalog No. 60045744 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-744 6 x 1 L
60-022-40 2.5 L
60-048-239 4 x 4 L
60-022-39 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-045-744 Supplier Solstice Supplier No. 2705556X1L
Add to Cart
Add to Cart

CHROMASOLV™ Plus, for HPLC,≥99.9%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20°C (literature)
Absorbance 0.01 at 360 nm
CAS Max % 100.0000%
Density 0.937 g/mL at 25 °C (literature)
Quantity 6 x 1 L
Flash Point 64°C
Packaging Glass Bottle
Linear Formula CH3CH2COCH2CH3
Beilstein 1737614
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide
Boiling Point 164°C to 166°C
Formula Weight 87.12
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material N,N-Dimethylacetamide, CHROMASOLV™ Plus, for HPLC, ≥99.9%
Show More Show Less
ShelfLife 720 days from date of manufacture
EINECSNumber 204-826-4
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.