Learn More
Sigma Aldrich Fine Chemicals Biosciences N,N-Dimethylacetamide | 127-19-5 | MFCD00008686 | 2 L
Catalog No. NC9072012
Encompass
N,N-Dimethylacetamide | Purity: ≥99.9% | Mol Wt: 87.12 | 127-19-5 | MFCD00008686 | 2 L
Catalog No. NC9072012 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2705552L
N,N-Dimethylacetamide | Purity: ≥99.9% | Mol Wt: 87.12 | 127-19-5 | MFCD00008686 | 2 L