Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DMAC, Dimethylacetamide, GC Headspace Grade, Thermo Scientific™

Catalog No. d1601 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D1601 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1601
Only null left
Add to Cart

DMAC processed to high purity for accurate and repeatable determination of trace levels of residual solvents.

  • Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • This product is UV Absorbance Tested to ensure low organic contamination
  • Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Absorbance 0.300 AU Maximum (275nm), 0.080 AU Maximum (300nm), 0.005 AU Maximum (350nm)
Color Colorless
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Titratable Acid 0.001 mEq/g Maximum
Water 0.03% Maximum
Formula Weight 87.12
Percent Purity ≥99.9%
Grade GC Headspace
Chemical Name or Material DMAC, Dimethylacetamide
Show More Show Less
Recommended Storage Room temperature, packaged under inert atmosphere
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.