Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylacrylamide

Catalog No. 274135500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-413-5500ML 500 mL
27-413-5100ML 100 mL
27-413-55ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 27-413-5500ML Supplier Sigma Aldrich Supplier No. 274135500ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N-Dimethylacrylamide (DMA) is a nonionic acrylic monomer that can make swellable polymeric particles.DMA is used in the synthesis of photoresponsive polymers. It is used in the synthesis of copolymers bygrafting polysiloxane on the side chain of DMA by homopolymerization orgrafting cellulose to form cellulose polymers andcopolymerization of methyl methacrylate with DMA orcopolymerization of DMA with styrene and butadiene

Specifications

Density 0.962 g/mL (at 25°C)
Boiling Point 80°C to 81°C (20 mmHg)
Molecular Formula C5H9NO
Refractive Index n20/D 1.473 (literature)
Linear Formula CH2=CHCON(CH3)2
Quantity 500 mL
MDL Number MFCD00008626
Synonym 2-Propenamide
Molecular Weight (g/mol) 99.13
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.