Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylallylamine 98.0+%, TCI America™
SDP

Catalog No. D116025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D116025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D116025ML Supplier TCI America Supplier No. D116025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An amine-based building block useful for chemical synthesis.

Chemical Identifiers

CAS 2155-94-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00038234
InChI Key GBCKRQRXNXQQPW-UHFFFAOYSA-N
Synonym n,n-dimethylallylamine, allyldimethylamine, dimethylallylamine, 2-propen-1-amine, n,n-dimethyl, 1-dimethylamino-2-propene, n-allyl-n,n-dimethylamine, n-allyldimethylamine, dimethyl prop-2-en-1-yl amine, allylamine, n,n-dimethyl, ch2=chch2n ch3 2
PubChem CID 75082
IUPAC Name dimethyl(prop-2-en-1-yl)amine
SMILES CN(C)CC=C

Specifications

Color Colorless
Boiling Point 64°C
Quantity 25 mL
UN Number 2733
Formula Weight 85.15
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material N,N-Dimethylallylamine
EINECSNumber (2)-2380
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.