Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (N,N-Dimethylaminopropyl)Trimethoxysilane

Catalog No. 53928725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-928-725ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 53-928-725ML Supplier Sigma Aldrich Supplier No. 53928725ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(N,N-Dimethylaminopropyl)Trimethoxysilane

Specifications

CAS 42437-96-7
Melting Point 344°C to 350°C
Molecular Formula C7H13NO -+ HCl
Linear Formula C7H13NO -+ HCl
Refractive Index n20/D 1.416
MDL Number MFCD00270837
Quantity 5 g
Synonym (R)-(-)-3-Hydroxyquinuclidine hydrochloride
Molecular Weight (g/mol) 163.65 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.