Learn More
N,N-Dimethylaniline, 99%
Description
N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler's ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler′s ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.
Solubility
Miscible with water, ethanol, ether, acetone, chloroform, benzene and alkaline.
Notes
Incompatible with strong oxidizing agents, Strong acids, acid chlorides, acid anhydrides, chloroformates and halogens.
Specifications
Specifications
| Melting Point | 2°C |
| Density | 0.956 |
| pH | 7.4 |
| Boiling Point | 193°C |
| Flash Point | 62°C (143°F) |
| Odor | Characteristic |
| Assay Percent Range | 99% |
| Refractive Index | 1.558 |
| Quantity | 10,000 mL |
| UN Number | UN2253 |
| Show More |
Safety and Handling
| Hazard Category | H227-H301+H311+H331-H351 |
| Hazard Statement | GHS H Statement H301-H311-H331-H351-H227 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Suspected of causing cancer. Combustible liquid. |
| Precautionary Statement | P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N,N-DIMETHYLANILINE |
| EINECSNumber | 204-493-5 |
| RTECSNumber | BX4725000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.