Learn More
eMolecules​ 98-94-2 | N,N-Dimethylcyclohexanamine | Oakwood Chemical | MFCD00003844 | 127.231 | C8H17N | 99.000 | CN(C)C1CCCCC1 | 25g | 571107366
Catalog No. NC2227676 Shop All eMolecules Inc. Products
Encompass_Preferred
N,N-Dimethylcyclohexanamine | Oakwood Chemical | 98-94-2 | MFCD00003844 | 127.231 | C8H17N | 99.000 | CN(C)C1CCCCC1 | 25g | 571107366
This item is not returnable. View return policy
Catalog No. NC2227676 Supplier eMolecules​ Supplier No. 571107366
N,N-Dimethylcyclohexanamine | Oakwood Chemical | 98-94-2 | MFCD00003844 | 127.231 | C8H17N | 99.000 | CN(C)C1CCCCC1 | 25g | 571107366