Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylcyclohexylamine

Catalog No. 2906291L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-062-91L 1 L
29-062-925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-062-91L Supplier Sigma Aldrich Supplier No. 2906291L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sampling and subsequent analytical determination of airborne n,n-dimethylcyclohexylamine has been reported.n,n-dimethylcyclohexylamine has been used:
  • As switchable hydrophilicity solvent (shs) for the extraction of lipids from freeze-dried samples of botryococcus braunii microalgae for biofuel production
  • As catalyst in three-component organocatalyzed strecker reaction on water

Specifications

CAS 98-94-2
Density 0.849 g/mL (at 25°C (literature))
Boiling Point 158°C to 159°C (lit.)
Molecular Formula C8H17N
Refractive Index n20/D 1.454 (literature)
Linear Formula C6H11N(CH3)2
MDL Number MFCD00003844
Quantity 1 L
Synonym Cyclohexyldimethylamine; Dimethylaminocyclohexane
Molecular Weight (g/mol) 127.23
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.