Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethyldipropylenetriamine

Catalog No. 550019500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-001-9500ML 500 mL
55-001-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-001-9500ML Supplier Sigma Aldrich Supplier No. 550019500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dimethyldipropylenetriamine is an aliphatic acyclic amine that is commonly used to develop polymeric system for gene delivery.n,n-dimethyldipropylenetriamine (dp) may be used to:
  • Prepare poly(ethylene oxide)-b-poly(3-caprolactone-g-dp) [peo-b-p(cl-g-dp)], a biodegradable amphiphilic polycationic copolymer
  • Develop a second generation aminoglycoside 6α-n-acetyltransferase (aac(6α) inhibitor
  • Prepare a hydrophilic gemcitabine conjugated cationic copolymer, which can be employed in the treatment pancreatic cancer.product of arkema inc

Specifications

CAS 10563-29-8
Density 0.883 g/mL (at 25°C (literature))
Boiling Point 220°C (lit.)
Molecular Formula C8H21N3
Refractive Index n20/D 1.4630 (literature)
Linear Formula (CH3)2 N(CH2)3 NH(CH2)3 NH
Quantity 500 mL
MDL Number MFCD00082192
Synonym DMAPAPA
Molecular Weight (g/mol) 159.27
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.