Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N'-Dimethylethylenediamine

Catalog No. D1578055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1578055G 5 g
D157805500G 500 g
D157805100G 100 g
D15780525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. D1578055G Supplier Sigma Aldrich Supplier No. D1578055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,nα-dimethylethylenediamine can be used as a ligand in the following processes:
  • Copper-catalyzed cyanations of aryl bromides using potassium hexacyanoferrate(ii) to form the corresponding benzonitriles
  • Copper-catalyzed tandem reaction of carbodiimide and isocyanoacetate to form benzoimidazo[1,5-α]imidazoles
  • Aminothiolation of 1,1-dibromoalkene to form n-fused heterocycles
  • In combination with ferrous chloride, for the n-arylation of nh-sulfoximines with aryl iodides to form n-arylsulfoximines
  • It can catalyze the direct c-h arylation of unactivated benzene in the presence of potassium tert-butoxide to form the corresponding biaryl

Specifications

CAS 110-70-3
Density 0.819 g/mL (at 20°C (literature))
Boiling Point 119°C (lit.)
Molecular Formula C4H12N2
Refractive Index n20/D 1.431 (literature)
Linear Formula CH3NHCH2CH2NHCH3
MDL Number MFCD00008290
Quantity 5 g
Synonym DMEDA; 1,2-Bis(methylamino)ethane
Molecular Weight (g/mol) 88.15
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.