Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dimethylethylenediamine, tech., 85%

Catalog No. AC116190050
Change view
Click to view available options
Quantity:
5 mL
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116190050 5 mL Glass bottle
AC116190250 25 g Glass bottle
AC116191000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC116190050 Supplier Thermo Scientific Chemicals Supplier No. 116190050

CAS: 110-70-3 | C4H12N2 | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-70-3
InChI Key KVKFRMCSXWQSNT-UHFFFAOYSA-N
Synonym n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine
PubChem CID 8070
IUPAC Name N,N'-dimethylethane-1,2-diamine
SMILES CNCCNC

Specifications

Color Colorless to Yellow
Density 0.8190g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C4H12N2
Linear Formula CH3NHCH2CH2NHCH3
Quantity 5 mL
Beilstein 04, 250
Molecular Weight (g/mol) 88.154
Formula Weight 88.15
Percent Purity ≥80%
Grade Technical
Physical Form Liquid
Chemical Name or Material N, N';-Dimethylethylenediamine, tech., 85%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-793-3
RTECSNumber KV4250000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.