Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Dimethylethylenediamine, tech., 85%

Catalog No. AC116191000
Change view
Click to view available options
Quantity:
5 mL
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116191000 100 g Glass bottle
AC116190250 25 g Glass bottle
AC116190050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC116191000 Supplier Thermo Scientific Chemicals Supplier No. 116191000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 110-70-3 | C4H12N2 | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-70-3
InChI Key KVKFRMCSXWQSNT-UHFFFAOYSA-N
Synonym n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine
PubChem CID 8070
IUPAC Name N,N'-dimethylethane-1,2-diamine
SMILES CNCCNC

Specifications

Color Colorless to Yellow
Density 0.8190g/mL
Boiling Point 119.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C4H12N2
Refractive Index 1.4295
Linear Formula CH3NHCH2CH2NHCH3
Quantity 100 g
Beilstein 04, 250
Specific Gravity 0.819
Solubility Information Solubility in water: miscible. Other solubilities: soluble in organic solvents
Molecular Weight (g/mol) 88.15
Formula Weight 88.15
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material N,N′-Dimethylethylenediamine, tech., 85%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse
EINECSNumber 203-793-3
RTECSNumber KV4250000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.