Learn More
Sigma Aldrich Fine Chemicals Biosciences N,N-Dimethylformamide, 68-12-2, MFCD00003284, 2 L
Catalog No. NC0761562
Encompass
Linear Formula: HCON(CH3)2, Molecular Weight: 73.09, for HPLC, ≥99.9%, Synonym: DMF, NSC 5356. Solvent for many hydrophobic organic compounds.
Catalog No. NC0761562 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2705472L
Linear Formula: HCON(CH3)2, Molecular Weight: 73.09, for HPLC, ≥99.9%, Synonym: DMF, NSC 5356. Solvent for many hydrophobic organic compounds.