Learn More
Sigma Aldrich Fine Chemicals Biosciences N,N-Dimethylformamide anhydrous, 68-12-2, MFCD00003284, 100mL
Catalog No. 501657133
Encompass
Linear Formula HCON(CH3)2, 99.8%, Molecular Weight 73.09, bp 153 °C (lit.), mp −61 °C (lit.), Synonym: DMF, NSC 5356
Catalog No. 50-165-7133 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 227056100ML
Linear Formula HCON(CH3)2, 99.8%, Molecular Weight 73.09, bp 153 °C (lit.), mp −61 °C (lit.), Synonym: DMF, NSC 5356