Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, 99.8%, Thermo Scientific™

Catalog No. AC326872500
Catalog No. AC326872500 Supplier Thermo Scientific Chemicals Supplier No. 326872500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61.0°C
Density 0.9450g/mL
Flash Point 58°C
Refractive Index 1.4290 to 1.4320
Specific Gravity 0.945
Linear Formula HCON(CH3)2
Viscosity 0.8 mPa.s (20°C)
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Boiling Point 153.0°C
Formula Weight 73.09
Grade Extra Dry
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.