Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide,≥99.99995% (trace metals analysis), Solstice TraceSELECT™,

Catalog No. 60047444 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-444 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-444 Supplier Solstice Supplier No. 727811L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceSELECT™, for inorganic trace analysis,≥99.99995% (trace metals analysis)

  • High purity solvents for speciation analysis by LC-ICP-MS

  • Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance

  • The blank values for metal traces in these solvents are in the ppb range or lower.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Density 0.947-0.949g/mL at 20°C, 0.944g/mL
Quantity 1 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Packaging Poly Bottle
Vapor Pressure 2.7mmHg (20°C)
Linear Formula HCON(CH3)2
UN Number UN2265
Beilstein 605365
Formula Weight 73.09g/mol
Percent Purity ≥99.99995% (trace metals analysis)
Grade TraceSELECT
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Statement P201-P280-P305 + P351 + P338-P308 + P313
DOTInformation 3/PGIII
RTECSNumber 200-679-5
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.