Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, 99%

Catalog No. AAA135470F
Change view
Click to view available options
Quantity:
10,000 mL
250 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA135470F 2500 mL
AAA13547AK 250 mL
AAA135470D 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA135470F Supplier Thermo Scientific Chemicals Supplier No. A135470F
  • $130.10
    $116.65 / Each of 1

    Save $13.45

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

CAS: 68-12-2 | C3H7NO | 73.10 g/mol

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

Solubility
Miscible with water and most organic solvents.

Notes
Reaction with sodium hydride gives exothermic decomposition at low temperatures. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61°C
Density 0.944
pH 7
Boiling Point 153°C
Flash Point 57°C (135°F)
Odor Amine-like
Refractive Index 1.431
Quantity 2500 mL
UN Number UN2265
Beilstein 605365
Merck Index 14,3243
Solubility Information Miscible with water and most organic solvents.
Formula Weight 73.09
Percent Purity 99%
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Category H226-H312+H332-H319-H335-H336-H350-H360D
Hazard Statement GHS H Statement
H360-H226-H312-H332-H319
May damage fertility or the unborn child.
Flammable liquid and vapour.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N,N-DIMETHYLFORMAMIDE
EINECSNumber 200-679-5
RTECSNumber LQ2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.