Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, ACS, 99.8%, Spectrum™ Chemical
SDP

Catalog No. 18611875 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-875 100 mL Amber Glass Bottle
18-611-878 500 mL Amber Glass Bottle
18-611-876 1 L Amber Glass Bottle
18-611-877 4 L Amber Glass Bottle
18-611-873 20 L Steel Can
18-611-874 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-875 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1030100ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68-12-2
Molecular Weight (g/mol) 73.09

Specifications

CAS Max % 1
Quantity 100 mL
Identification To Pass Test
Packaging Amber Glass Bottle
Residue after Evaporation 5e-05
Titratable Acid 0.0005 meq/g
Titratable Base 0.003 meq/g
Water 0.0015
Boiling Point 153°C
Percent Purity 0.998
Grade Reagent
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.