Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, for HPLC, GC and spectrophotometry, >99.9%, Solstice

Catalog No. 6001009 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-010-09 6 x 1 L Glass Bottle
60-010-08 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-010-09 Supplier Solstice Supplier No. 0761L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Absorbance 0.3 AU max. at 275 nm (UV), 0.05 AU max. at 300 nm (UV), 0.005 AU max. at 350 nm (UV), 0.005 AU max. at 400 nm (UV)
CAS Max % 100.0000%
Density 0.949 g/cm3
Quantity 6 x 1 L
Flash Point 58°C
Packaging Glass Bottle
UN Number UN2265
Water 0.03% max.
Beilstein 605365
Boiling Point 153°C
Solubility Information Miscible in Water at 20°C
Formula Weight 73.10
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material N,N-Dimethylformamide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: N,N-Dimethylformamide
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.