Learn More
Supply Solutions Burdick & Jackson - N,N-Dimethylformamide, BioSyn. Package Size: 200L. Container Type:SSPDS. Suggested Applications: Biosynthesis, Applications requiring low water content
Catalog No. 50877150
Encompass_Preferred
N,N-Dimethylformamide, BioSyn. Suggested Application: Biosynthesis, Applications requiring low water content. Package Size: 20L. Container type is steel NOWPAK. CAS Number: 68-12-2. Hazard Class: 3. Package Group: III. Molecular Weight: 73.10. Density (g/mL) at 20C: 0.9487. Boiling point (C): 153.0. Freezing point (C): -60.4. Flashpoint (C): 58. Manufacturer Name: Burdick and Jackson. Manufacturer Part Number: BN075-20.; Size: 200L
Catalog No. 50-877-150 Supplier Supply Solutions Supplier No. BJ BC075200
Add to Cart
Add to Cart
Request a Quote
N,N-Dimethylformamide, BioSyn. Suggested Application: Biosynthesis, Applications requiring low water content. Package Size: 20L. Container type is steel NOWPAK. CAS Number: 68-12-2. Hazard Class: 3. Package Group: III. Molecular Weight: 73.10. Density (g/mL) at 20C: 0.9487. Boiling point (C): 153.0. Freezing point (C): -60.4. Flashpoint (C): 58. Manufacturer Name: Burdick and Jackson. Manufacturer Part Number: BN075-20.; Size: 200L