Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide CHROMASOLV™ Plus, for HPLC, ≥99.9%, Solstice

Catalog No. 6000997 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-009-97 1 L
60-009-98 2.5 L
60-010-00 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 60-009-97 Supplier Solstice Supplier No. 2705471L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Solvent for many hydrophobic organic compounds.

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61°C
Density 0.944g/mL
Quantity 1 L
Refractive Index n20/D 1.430
Linear Formula HCON(CH3)2
Vapor Pressure 2.7mmHg (20 °C)
Beilstein 605365
Boiling Point 153°C
Formula Weight 73.09g/mol
Percent Purity ≥99.9%
Grade glass distillation
Chemical Name or Material N,N-Dimethylformamide, for HPLC
Show More Show Less
Hazard Statement P201-P280-P305 + P351 + P338-P308 + P313
Precautionary Statement H226-H312 + H332-H319-H360D
EINECSNumber 200-679-5
RTECSNumber LQ2100000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.