Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylformamide di-tert-butyl acetal

Catalog No. 4026310MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-263-10MLF 10 mL
40-263-50MLF 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-263-10MLF Supplier Sigma Aldrich Supplier No. 4026310MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dimethylformamide di-tert-butyl acetal may be used in the synthesis of following:
  • (s)-2-(1-tert-butoxycarbonyl-2-{4-[2-(5-methyl-2-phenyloxazol-4-yl)ethoxy]phenyl}ethylamino)benzoic acid methyl ester
  • 3-o-tert-butylmorphine
  • Tert-butylesters of pyrrole- and indolecarboxylic acidsreview; synthesis of tert-butyl esters

Specifications

CAS 36805-97-7
Density 0.848 g/mL (at 25°C (literature))
Boiling Point 56°C to 57°C (8 mmHg)
Molecular Formula C11H25NO2
Refractive Index n20/D 1.413 (literature)
Linear Formula (CH3)2 NCH[OC(CH3)3]2
MDL Number MFCD00015002
Quantity 10 mL
Synonym 1,1-Di-tert-butoxy-N,N-dimethylmethylamine; 1,1-Di-tert-butoxytrimethylamine
Molecular Weight (g/mol) 203.32
Percent Purity ≥90% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.