Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide di-tert-butyl acetal, ≥90.0%, MilliporeSigma™ Supelco™

Catalog No. 111018236 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8236 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8236 Supplier MilliporeSigma Supelco Supplier No. 39500525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N-Dimethylformamide di-tert-butyl acetal is an derivatizing reagent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00015002
Synonym 1,1-Di-tert-butoxy-N,N-dimethylmethylamine; 1,1-Di-tert-butoxytrimethylamine

Specifications

Density 0.848 g/mL (at 25°C)
CAS 36805-97-7
Boiling Point 56°C to 57°C (8 mmHg)
Flash Point 35°C - Closed Cup
Packaging Ampule
Linear Formula (CH3)2 NCH[OC(CH3)3]2
Quantity 25 mL
Formula Weight 203.32
Percent Purity ≥90.0%
Grade For GC derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.