Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylformamide diethyl acetal

Catalog No. 14277850G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-277-850G 50 g
14-277-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-277-850G Supplier Sigma Aldrich Supplier No. 14277850G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N-Dimethylformamide diethyl acetal undergoes Me3SiCl-mediated three-component coupling reaction with alkyne to yield 2,3,4,5-tetrasubstituted pyridine derivatives.N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.

Specifications

CAS 1188-33-6
Density 0.859 g/mL (at 25°C (literature))
Boiling Point 130°C to 133°C (lit.)
Molecular Formula C7H17NO2
Refractive Index n20/D 1.400 (literature)
Linear Formula (CH3)2 NCH(OC2H5)2
MDL Number MFCD00009227
Quantity 50 g
Synonym 1,1-Diethoxy-N,N-dimethylmethylamine; 1,1-Diethoxytrimethylamine
Molecular Weight (g/mol) 147.22
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.