Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide dimethyl acetal, >95%, MilliporeSigma™ Supelco™

Catalog No. 111019201 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9201 5 mL
11-101-8746 10 x 1 mL
11-101-8564 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-9201 Supplier MilliporeSigma Supelco Supplier No. 3949635ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Esterate M (N,N-Dimethylformamide dimethyl acetal, DMFDMA) is an methylating reagent. It has been used in methyl esterification of carboxylic acid and also in one-step derivatization of amino acids into N,N-Dimethylaminomethylene methyl esters.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00008482
Synonym 1,1-Dimethoxy-N,N-dimethylmethylamine; 1,1-Dimethoxytrimethylamine

Specifications

CAS 4637-24-5
Density 0.897 g/mL (at 25°C)
Boiling Point 102°C to 103°C (720 mmHg)
Flash Point 5°C - Closed Cup
Packaging Ampule
Linear Formula (CH3)2 NCH(OCH3)2
Quantity 5 mL
Formula Weight 119.16
Percent Purity >95%
Grade For GC derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.