Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide dipropyl acetal, ≥97.5%, MilliporeSigma™ Supelco™

Catalog No. 111018596 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8596 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8596 Supplier MilliporeSigma Supelco Supplier No. 39499820ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N-Dimethylformamide dipropyl acetal is an derivatizing reagent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00009374
Synonym 1,1-Dipropoxy-N,N-dimethylmethylamine; 1,1-Dipropoxytrimethylamine

Specifications

Density 0.854 g/mL (at 25°C)
CAS 6006-65-1
Boiling Point 67°C to 68°C (14 mmHg)
Flash Point 46°C - Closed Cup
Packaging Ampule
Linear Formula (CH3)2 NCH(OCH2 CH2 CH3)2
Quantity 20 mL
Formula Weight 175.27
Percent Purity ≥97.5%
Grade For GC derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.