Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, Solstice

Catalog No. 60090394 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Returnable NOWPAK™ II
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-394 20 L Returnable NOWPAK™ II
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-394 Supplier Solstice Supplier No. BN07520
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BioSyn™, for biosynthesis and applications requiring low water content, >99.9%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
PubChem CID 6228
ChEBI CHEBI:17741

Specifications

Quantity 20 L
Flash Point 58 °C
Packaging Returnable NOWPAK™ II
Physical Form Liquid
Grade Low Water
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.