Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, Solstice™

Catalog No. 60090239 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-239 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-239 Supplier Solstice Supplier No. 517816X1L/DEL

missing translation for 'validMainframeMsg'

GC-Headspace tested, ≥99.9%

  • Very low levels of non-volatile impurities
  • TRUSTED_SUSTAINABILITY

    Chemical Identifiers

    CAS 68-12-2
    PubChem CID 6228
    ChEBI CHEBI:17741

    Specifications

    Quantity 6 x 1 L
    Flash Point 58 °C
    Packaging Glass Bottle
    Physical Form Liquid
    Percent Purity min. 99.9 %
    Recommended Storage Room Temperature
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.