Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylglycine 98.0+%, TCI America™
SDP

Catalog No. D35855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35855G 5 g
D358525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35855G Supplier TCI America Supplier No. D35855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1118-68-9
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00004283
InChI Key FFDGPVCHZBVARC-UHFFFAOYSA-N
Synonym n,n-dimethylglycine, dimethylglycine, 2-dimethylamino acetic acid, dimethylamino acetic acid, n,n-dimethylaminoacetic acid, glycine, n,n-dimethyl, n-methylsarcosine, n,n-dimethyl glycine, unii-7797m4cppa, dmg
PubChem CID 673
ChEBI CHEBI:17724
IUPAC Name 2-(dimethylamino)acetic acid
SMILES CN(C)CC(=O)O

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 5 g
Formula Weight 103.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethylglycine
RTECSNumber MB9865000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.