Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, N,N-Dimethylglycine, 97%, [1118-68-9], D15436 103.12 C4H9NO2

Catalog No. 501409039
Encompass_Preferred

Compound: N,N-Dimethylglycine

CAS#: 1118-68-9

MDL#: not listed

Background: A dimethylamino-substituted acetic acid used as a common building block in pharmaceutical research. The tertiary amine is a common solubilizing motif, while the carboxylic acid group allows for amide coupling.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-140-9039 Supplier Frontier Specialty Chemicals Supplier No. D154361G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: N,N-Dimethylglycine

CAS#: 1118-68-9

MDL#: not listed

Background: A dimethylamino-substituted acetic acid used as a common building block in pharmaceutical research. The tertiary amine is a common solubilizing motif, while the carboxylic acid group allows for amide coupling.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.