Learn More
Cayman Chemical N,N-Dimethylglycine (hydrochloride), Purity: ≥98%, CAS: 2491-06-7, 10 g
SDP
Catalog No. 502949613
Encompass_Preferred
A synthetic intermediate; has been used in the synthesis of water-soluble prodrug forms of vitamin E and vitamin K1
Catalog No. 50-294-9613 Supplier Cayman Chemical Supplier No. 3383010 G
A synthetic intermediate; has been used in the synthesis of water-soluble prodrug forms of vitamin E and vitamin K1