Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylhexylamine

Catalog No. 30810225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-810-225G 25 g
30-810-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-810-225G Supplier Sigma Aldrich Supplier No. 30810225G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The free salinomycin (sali), that has selective toxicity to cancer stem cells (cscs), was encapsulated into the nanoparticle along with the additive n,n-dimethylhexylamine.n,n-dimethylhexylamine was used as ion-pairing agent to study the problem of poor retention of uridine diphosphate-linked intermediates on reverse phase media
  • It was also used in the determining the five monophosphate nucleotides (cytidine 5α-monophosphate, uridine 5α-monophosphate, adenosine 5α-monophosphate, inosine 5α-monophosphate and guanosine 5α-monophosphate) in baby foods

Specifications

CAS 4385-04-0
Density 0.744 g/mL (at 25°C (literature))
Boiling Point 146°C to 150°C (lit.)
Molecular Formula C8H19N
Refractive Index n20/D 1.414 (literature)
Linear Formula CH3(CH2)4 CH2N(CH3)2
MDL Number MFCD00009522
Quantity 25 g
Molecular Weight (g/mol) 129.24
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.