Learn More
Medchemexpress LLC N,N'-dimethylhydrazine dihydrochloride | 306-37-6 | MFCD00012522 | 98.0% | 133.02 g·mol⁻¹ | C2H10Cl2N2 | 5 G
SDP
Catalog No. 5000284397
Encompass_Preferred
N,N'-Dimethylhydrazine dihydrochloride is the dihydrochloride salt of 1,2-dimethylhydrazine, used as a research reagent to induce colon tumors in rodent models. Supplied as a high-purity solid for laboratory use, it requires careful handling, sealed storage, and appropriate safety controls due to its carcinogenic activity.
  • Research reagent for induction of colon tumors in rodents.
  • High purity suitable for experimental protocols.
  • Solid dihydrochloride salt, soluble in water.
  • Available in small research pack sizes for laboratory studies.
  • Store at 4°C, sealed, away from moisture and light.
Catalog No. 50-002-84397 Supplier Medchemexpress LLC Supplier No. HYW4604075G
N,N'-Dimethylhydrazine dihydrochloride is the dihydrochloride salt of 1,2-dimethylhydrazine, used as a research reagent to induce colon tumors in rodent models. Supplied as a high-purity solid for laboratory use, it requires careful handling, sealed storage, and appropriate safety controls due to its carcinogenic activity.
  • Research reagent for induction of colon tumors in rodents.
  • High purity suitable for experimental protocols.
  • Solid dihydrochloride salt, soluble in water.
  • Available in small research pack sizes for laboratory studies.
  • Store at 4°C, sealed, away from moisture and light.