Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylisopropylamine

Catalog No. 550051100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-005-1100ML 100 mL
55-005-1500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55-005-1100ML Supplier Sigma Aldrich Supplier No. 550051100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dimethylisopropylamine may be used as an alkylating agent in the preparation of n-methylaniline via palladium catalyzed n-methylation of aniline under microwave condition
  • It may also be used to prepare a hypergolic ionic liquid by reacting with hydrazoic acid.product of arkema inc

Specifications

CAS 996-35-0
Density 0.715 g/mL (at 25°C (literature))
Boiling Point 65.5°C (752 mmHg)
Molecular Formula C5H13N
Refractive Index n20/D 1.3905 (literature)
Linear Formula (CH3)2 CHN(CH3)2
MDL Number MFCD00015014
Quantity 100 mL
Synonym DMIPA
Molecular Weight (g/mol) 87.16
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.