Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylmethyleneammonium Iodide 97.0+%, TCI America™
SDP

Catalog No. D164625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D164625G 25 g
D16465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D164625G Supplier TCI America Supplier No. D164625G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33797-51-2
Molecular Formula C3H8IN
Molecular Weight (g/mol) 185.008
MDL Number MFCD00011810
InChI Key VVDUZZGYBOWDSQ-UHFFFAOYSA-M
Synonym eschenmoser's salt, n,n-dimethylmethyleneiminium iodide, n-methyl-n-methylenemethanaminium iodide, dimethyl methylidene azanium iodide, n,n-dimethyl methyleneammonium iodide, dimethyl methylene ammonium iodide, methanaminium, n-methyl-n-methylene-, iodide, eschenmoser inverted exclamation mark s salt, eschenmoser salt, dimethyl methylene ammounium iodide
PubChem CID 2724133
IUPAC Name dimethyl(methylidene)azanium;iodide
SMILES C[N+](=C)C.[I-]

Specifications

Quantity 25 g
Formula Weight 185.01
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethylmethyleneammonium Iodide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.