Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyloctylamine, 97%

Catalog No. AC225290025
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225290025 2.5 L Glass bottle
AC225290250 25 mL Glass bottle
AC225295000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC225290025 Supplier Thermo Scientific Chemicals Supplier No. 225290025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 7378-99-6 | C10H23N | 157.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7378-99-6
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00009558
InChI Key UQKAOOAFEFCDGT-UHFFFAOYSA-N
Synonym n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd
PubChem CID 16224
IUPAC Name N,N-dimethyloctan-1-amine
SMILES CCCCCCCCN(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -57.0°C
Density 0.7650g/mL
Boiling Point 195.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (C8-chains) (GC)
Packaging Glass bottle
Refractive Index 1.4233 to 1.4253
Linear Formula CH3(CH2)7N(CH3)2
Quantity 2.5 L
Beilstein 04,I,386
Specific Gravity 0.765
Solubility Information Solubility in water: slightly soluble
Formula Weight 157.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyloctylamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contac
EINECSNumber 230-939-3
RTECSNumber RG8075000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.