Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyloxamic Acid 97.0+%, TCI America™
SDP

Catalog No. D42415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42415G 5 g
D42411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42415G Supplier TCI America Supplier No. D42415G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32833-96-8
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.104
MDL Number MFCD01971023
InChI Key YKFGLGXRUVEMNF-UHFFFAOYSA-N
Synonym N,N-Dimethyloxamidic Acid
PubChem CID 2760995
IUPAC Name 2-(dimethylamino)-2-oxoacetic acid
SMILES CN(C)C(=O)C(=O)O

Specifications

Melting Point 125°C
Color White
Quantity 5 g
Formula Weight 117.10
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dimethyloxamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.