Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylphosphoramic dichloride

Catalog No. 7962510ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-625-10ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-625-10ML Supplier Sigma Aldrich Supplier No. 7962510ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N-Dimethylphosphoramic dichloride

Specifications

CAS 677-43-0
Density 1.362 g/mL (at 20°C (literature))
Boiling Point 77°C to 79°C (11 mmHg)
Molecular Formula C2H6Cl2NOP
Refractive Index n20/D 1.464
Linear Formula (CH3)2 NP(O)Cl2
MDL Number MFCD00013896
Quantity 1 g
Synonym DMPADC; Dimethylamidophosphoric dichloride
Molecular Weight (g/mol) 161.95
Percent Purity ≥98.0% (AT)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.