Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N,N-Dimethylpropionamide, ≥98%

Click to view available options
Quantity:
25g
100g
500g
758-96-3
C5H11NO
101.149
MFCD00009301
MBHINSULENHCMF-UHFFFAOYSA-N
n,n-dimethylpropionamide,propanamide, n,n-dimethyl,dimethylamide of propionic acid,propionamide, n,n-dimethyl,propionamide, n,n-dimethyl,n,n-dimethyl propionamide,acmc-209p0o,propionic acid dimethylamide,4-04-00-00184 beilstein handbook reference,ksc494e1j
12965
N,N-dimethylpropanamide
CCC(=O)N(C)C
758-96-3
C5H11NO
101.149
MFCD00009301
MBHINSULENHCMF-UHFFFAOYSA-N
n,n-dimethylpropionamide,propanamide, n,n-dimethyl,dimethylamide of propionic acid,propionamide, n,n-dimethyl,propionamide, n,n-dimethyl,n,n-dimethyl propionamide,acmc-209p0o,propionic acid dimethylamide,4-04-00-00184 beilstein handbook reference,ksc494e1j
12965
N,N-dimethylpropanamide
CCC(=O)N(C)C
Applications
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.

Solubility
Soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Density 0.920g/mL
Boiling Point 174°C to 175°C (literature)
Quantity 100g
Melting Point -45°C (literature)
Refractive Index 1.4400
Color Colorless
Linear Formula CH3CH2CON(CH3)2
Beilstein 1740719
Formula Weight 101.15
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material N,N-Dimethylpropionamide
Show More Show Less

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UE3810000

TSCA : TSCA