Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N-Dimethylsulfamoyl chloride

Catalog No. D18625225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18625225G 25 g
D186252100G 100 g
D186252500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D18625225G Supplier Sigma Aldrich Supplier No. D18625225G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Applications of n,n -dimethylsulfamoyl chloride are:
  • It is widely used in the synthesis of a medicinally important class of compounds, sulfonamides
  • It is used in the synthesis of aminotetralin derived sulfamides as anticancer agents and acetylcholinesterase inhibitors
  • It can also be used in the esterification/amidation between carboxylic acids and equimolar amounts of alcohols/amines, as in the synthesis of coumaperine

Specifications

CAS 13360-57-1
Density 1.337 g/mL (at 25°C (literature))
Boiling Point 114°C (75 mmHg)
Molecular Formula C2H6ClNO2S
Refractive Index n20/D 1.452 (literature)
Linear Formula (CH3)2 NSO2Cl
MDL Number MFCD00007425
Quantity 25 g
Molecular Weight (g/mol) 143.59
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.