Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylthioformamide, MP Biomedicals

Catalog No. ICN224414 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224414 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224414 Supplier MP Biomedicals, Inc Supplier No. 0522441410

Chemical Identifiers

CAS 758-16-7
Molecular Formula C3H7NS
Molecular Weight (g/mol) 89.156
InChI Key SKECXRFZFFAANN-UHFFFAOYSA-N
Synonym n,n-dimethylthioformamide, dimethylthioformamide, methanethioamide, n,n-dimethyl, formamide, n,n-dimethylthio, nn-dimethylthioformamide, methanethioamide,n,n-dimethyl, formamide,n-dimethylthio, dimethylamino methanethial, n,n-dimethyl-thioformamide, methanethioamide, dimethyl
PubChem CID 69794
IUPAC Name N,N-dimethylmethanethioamide
SMILES CN(C)C=S

Specifications

Quantity 10 g
Formula Weight 89.16
Chemical Name or Material N, N-Dimethyl Thioformamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.