Learn More
Medchemexpress LLC N,N'-dimethyltrimethyleneurea | 7226-23-5 | 99.9% | 128.17 g/mol | C6H12N2O | 1000g
SDP
Catalog No. 5000270098
Encompass_Preferred
N N -Dimethyltrimethyleneurea is an intermediate N N -Dimethyltrimethyleneurea can be used in the preparation of naphthalocyanine compounds (near-infrared absorbing dyes)[1]
Catalog No. 50-002-70098 Supplier Medchemexpress LLC Supplier No. HYY01001000G
N N -Dimethyltrimethyleneurea is an intermediate N N -Dimethyltrimethyleneurea can be used in the preparation of naphthalocyanine compounds (near-infrared absorbing dyes)[1]