Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N-dimethyltryptamine N-oxide | 948-19-6 | ≥98.0% | 204.3 g/mol | C12H16N2O | 1 MG
SDP

Catalog No. 5000284233
Encompass_Preferred

N,N-dimethyltryptamine N-oxide is the N-oxide metabolite of N,N-dimethyltryptamine (DMT). Supplied as a research-grade reference standard, it is used in analytical and forensic laboratories for identification and quantification of DMT metabolites, and as an authenticated material for method development.

  • High purity: ≥98%.
  • Chemical formula: C12H16N2O.
  • Molecular weight: 204.3 g/mol.
  • CAS number: 948-19-6.
  • Intended for analytical and forensic research applications.
  • Typically provided as a crystalline solid for handling and storage.

Catalog No. 50-002-84233 Supplier Medchemexpress LLC Supplier No. HYW5763121MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N-dimethyltryptamine N-oxide is the N-oxide metabolite of N,N-dimethyltryptamine (DMT). Supplied as a research-grade reference standard, it is used in analytical and forensic laboratories for identification and quantification of DMT metabolites, and as an authenticated material for method development.

  • High purity: ≥98%.
  • Chemical formula: C12H16N2O.
  • Molecular weight: 204.3 g/mol.
  • CAS number: 948-19-6.
  • Intended for analytical and forensic research applications.
  • Typically provided as a crystalline solid for handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.