Learn More
Medchemexpress LLC N,N'-Dinitrosopiperazine | 140-79-4 | 99.94% | 144.13 | 1 ML
SDP
Catalog No. 5000349170
Encompass_Preferred
N,N'-Dinitrosopiperazine (DNP) is identified as a carcinogen with specificity for nasopharyngeal epithelium. It facilitates nasopharyngeal carcinoma (NPC) metastasis by regulating various signaling pathways, notably through the phosphorylation of LYRIC at serine 568. This compound is offered as a high-purity solution for research applications.
  • Carcinogen with specificity for nasopharyngeal epithelium
  • Facilitates NPC metastasis
  • Regulates multiple signaling pathways via protein phosphorylation
  • Targets LYRIC at serine 568
  • Supplied as a 10 mM solution in DMSO
  • High purity of 99.94%
Catalog No. 50-003-49170 Supplier Medchemexpress LLC Supplier No. HY13643310MM/1ML
N,N'-Dinitrosopiperazine (DNP) is identified as a carcinogen with specificity for nasopharyngeal epithelium. It facilitates nasopharyngeal carcinoma (NPC) metastasis by regulating various signaling pathways, notably through the phosphorylation of LYRIC at serine 568. This compound is offered as a high-purity solution for research applications.
  • Carcinogen with specificity for nasopharyngeal epithelium
  • Facilitates NPC metastasis
  • Regulates multiple signaling pathways via protein phosphorylation
  • Targets LYRIC at serine 568
  • Supplied as a 10 mM solution in DMSO
  • High purity of 99.94%