Learn More
Medchemexpress LLC N,N'-dinitrosopiperazine | 140-79-4 | MFCD00014615 | >95.0% | 144.13 | C4H8N4O2 | 5 MG
SDP
Catalog No. 5000221835
Encompass_Preferred
N,N'-dinitrosopiperazine is a small-molecule nitrosamine reagent used in biochemical and toxicological research. It is reported as a carcinogen with specificity for the nasopharyngeal epithelium and is supplied as a solid research chemical to be handled with appropriate laboratory safety precautions.
  • Small-molecule nitrosamine used for biochemical and toxicological research.
  • Documented carcinogenic activity with nasopharyngeal specificity in the literature.
  • Provided as a high-purity reagent suitable for analytical applications.
  • Available in small vial sizes appropriate for assay-scale use.
  • Store at 4°C and protect from light; in solution, store at -80°C for long-term stability.
Catalog No. 50-002-21835 Supplier Medchemexpress LLC Supplier No. HY1364335MG
N,N'-dinitrosopiperazine is a small-molecule nitrosamine reagent used in biochemical and toxicological research. It is reported as a carcinogen with specificity for the nasopharyngeal epithelium and is supplied as a solid research chemical to be handled with appropriate laboratory safety precautions.
  • Small-molecule nitrosamine used for biochemical and toxicological research.
  • Documented carcinogenic activity with nasopharyngeal specificity in the literature.
  • Provided as a high-purity reagent suitable for analytical applications.
  • Available in small vial sizes appropriate for assay-scale use.
  • Store at 4°C and protect from light; in solution, store at -80°C for long-term stability.